Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBSCR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022479.2 | NP_071924.1 | 5 | 1094 | ||
XM_011516467.2 | XP_011514769.1 | 5 | 1217 | |||
XM_011516469.2 | XP_011514771.1 | 5 | 1217 | |||
XM_017012521.1 | XP_016868010.1 | 5 | 1217 | |||
GenBank mRNA | AB078148.1 | - | 5 | 828 | ||
AF410457.1 | - | 5 | 828 | |||
AJ626726.1 | - | 5 | 828 | |||
AK091379.1 | - | 1 | 44 | |||
AK094958.1 | - | 5 | 762 | |||
AK126044.1 | - | 5 | 1230 | |||
AK290036.1 | - | 5 | 1097 | |||
BC067524.1 | - | 5 | 828 | 7002169 | ||
BC067525.1 | - | 5 | 828 | 7002172 | ||
BC069624.1 | - | 5 | 828 | 7262423 | ||
BC069628.1 | - | 5 | 828 | 7262435 | ||
BC069636.1 | - | 5 | 828 | 7262447 | ||
BC069645.1 | - | 5 | 828 | 7262459 | ||
BC069997.1 | - | 5 | 828 | 7002171 |