Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022481.5 | NP_071926.4 | 32, 33 | 4221 | ||
XM_005268498.1 | XP_005268555.1 | 32, 33 | 4125 | |||
XM_005268499.1 | XP_005268556.1 | 32, 33 | 4159 | |||
XM_006714792.1 | XP_006714855.1 | 31, 32 | 3972 | |||
XM_011537677.2 | XP_011535979.1 | 30, 31 | 3640 | |||
GenBank mRNA | AJ310567.1 | - | 31, 32 | 4171 | ||
AK001579.1 | - | 15, 16 | 2145 | |||
AK024718.1 | - | 2, 3 | 107 | |||
AK091385.1 | - | 31, 32 | 3887 | |||
AK290052.1 | - | 32, 33 | 4214 | |||
BC009968.2 | - | 6, 7 | 472 |