Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTMR14 SIRNA Assays ›
Gene Symbol:
MTMR14
Gene Name:
myotubularin related protein 14
Gene Aliases:
C3orf29
Chromosome Location:
Chr.3: 9649433 - 9702394 on Build GRCh38
UniGene ID:
Hs.475382
Species:
Human
siRNA ID
HSS127601
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTMR14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077525.2
NP_001070993.1
16
1494
NM_001077526.2
NP_001070994.1
16
1494
NM_022485.4
NP_071930.2
16
1494
XM_005265400.1
XP_005265457.1
15
1392
XM_011534012.1
XP_011532314.1
15
1263
XM_011534013.1
XP_011532315.1
15
1219
XM_017007034.1
XP_016862523.1
17
1664
XM_017007035.1
XP_016862524.1
16
1589
XM_017007036.1
XP_016862525.1
17
1663
XM_017007037.1
XP_016862526.1
16
1589
XM_017007038.1
XP_016862527.1
15
1392
XM_017007039.1
XP_016862528.1
17
1663
XM_017007040.1
XP_016862529.1
16
1589
XM_017007041.1
XP_016862530.1
14
1188
XM_017007042.1
XP_016862531.1
15
1392
XM_017007043.1
XP_016862532.1
15
1263
XM_017007044.1
XP_016862533.1
14
1310
XM_017007045.1
XP_016862534.1
14
1299
GenBank mRNA
AF289603.1
-
16
1457
AK026058.1
-
15
1061
AK074792.1
-
16
1465
AK298846.1
-
10
952
AL713695.1
-
16
1415
AY007098.1
-
13
902
AY545552.1
-
3
137
BC001674.1
-
16
1399
2961594
BC025952.1
-
16
1444
4876556
BC035690.1
-
16
1406
5755887
BC050626.1
-
16
1417
4416416
BF796406.1
-
1
17
DQ630520.1
-
16
1343
Back To Top
More Information
Panther Classification:
Molecular Function -
hydrolase
phosphatase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylinositol biosynthetic process
macroautophagy
peptidyl-tyrosine dephosphorylation
phosphatidylinositol-3-phosphatase activity
protein serine/threonine phosphatase activity
protein tyrosine phosphatase activity
protein binding
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0