Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022569.2 | NP_072091.1 | 13, 14 | 3186 | ||
XM_017008545.1 | XP_016864034.1 | 12, 13 | 2184 | |||
XM_017008546.1 | XP_016864035.1 | 11, 12 | 1490 | |||
GenBank mRNA | AB036429.1 | - | 13, 14 | 3186 | ||
AK289670.1 | - | 13, 14 | 3164 | |||
BC113075.2 | - | 12, 13 | 1873 |