Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022659.3 | NP_073150.2 | 14 | 1696 | ||
GenBank mRNA | AK021562.1 | - | 7 | 735 | ||
AK304372.1 | - | 13 | 1344 | |||
AY700779.1 | - | 6 | 624 | |||
BC069665.1 | - | 7 | 713 | |||
BC069726.1 | - | 7 | 716 | |||
BC069747.1 | - | 7 | 715 | |||
BC069768.1 | - | 7 | 716 | |||
BC074794.2 | - | 6 | 652 | |||
BC113478.1 | - | 6 | 680 | |||
BC113504.1 | - | 6 | 680 |