Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033055.2 | NP_149044.2 | 11, 12 | 1255 | ||
XM_005271118.1 | XP_005271175.1 | 12, 13 | 1531 | |||
XM_011541957.1 | XP_011540259.1 | 10, 11 | 1296 | |||
XM_017002084.1 | XP_016857573.1 | 12, 13 | 1498 | |||
GenBank mRNA | AF427492.1 | - | 11, 12 | 1255 | ||
AK057172.1 | - | 11, 12 | 1391 | |||
AK075208.1 | - | 5, 6 | 456 | |||
AK096669.1 | - | 8, 9 | 906 | |||
AL137576.1 | - | 3, 4 | 349 | |||
BC064409.1 | - | 8, 9 | 912 | |||
BC107768.1 | - | 8, 9 | 1004 |