Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282949.2 | NP_001269878.1 | 5, 6 | 689 | ||
NM_001282951.2 | NP_001269880.1 | 4, 5 | 702 | |||
NM_001282952.2 | NP_001269881.1 | 5, 6 | 734 | |||
NM_022730.3 | NP_073567.1 | 6, 7 | 810 | |||
XM_017004716.1 | XP_016860205.1 | 6, 7 | 867 | |||
XM_017004717.1 | XP_016860206.1 | 5, 6 | 768 | |||
GenBank mRNA | AK022674.1 | - | 6, 7 | 741 | ||
AK024273.1 | - | 5, 6 | 683 | |||
AK098334.1 | - | 3, 4 | 1147 | |||
AK124133.1 | - | 6, 7 | 809 | |||
AK222879.1 | - | 6, 7 | 748 | |||
AK307486.1 | - | 5, 6 | 627 | |||
BC010739.1 | - | 3, 4 | 345 |