Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022761.2 | NP_073598.1 | 1, 2 | 388 | ||
XM_017018129.1 | XP_016873618.1 | 1, 2 | 109 | |||
GenBank mRNA | AJ250229.1 | - | 1, 2 | 86 | ||
AK027152.1 | - | 1, 2 | 71 | |||
BC020628.1 | - | 1, 2 | 107 | 4656696 | ||
CR457378.1 | - | 1, 2 | 51 |