Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288794.1 | NP_001275723.1 | 12 | 1549 | ||
NM_001288795.1 | NP_001275724.1 | 11 | 1398 | |||
NM_022762.4 | NP_073599.2 | 11 | 1409 | |||
XM_005265969.3 | XP_005266026.1 | 10 | 1229 | |||
XM_005265971.4 | XP_005266028.1 | 11 | 1976 | |||
XM_005265972.3 | XP_005266029.1 | 10 | 1356 | |||
XM_017009727.1 | XP_016865216.1 | 10 | 2020 | |||
XM_017009728.1 | XP_016865217.1 | 10 | 1687 | |||
GenBank mRNA | AK025971.1 | - | 8 | 954 | ||
AK093481.1 | - | 11 | 1350 | |||
AK093622.1 | - | 3 | 1866 | |||
AK094065.1 | - | 8 | 1556 | |||
AK122692.1 | - | 6 | 2272 | |||
AK125337.1 | - | 12 | 1496 | |||
AL137381.1 | - | 5 | 522 | |||
AL713670.1 | - | 10 | 1272 | |||
AY359092.1 | - | 8 | 2267 | |||
BC009911.2 | - | 11 | 1343 | 2820100 | ||
CA440193.1 | - | 1 | 570 | |||
DQ494789.1 | - | 11 | 1343 |