Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135095.1 | NP_001128567.1 | 22, 23 | 2898 | ||
NM_022763.3 | NP_073600.3 | 22, 23 | 2971 | |||
XM_017007062.1 | XP_016862551.1 | 21, 22 | 2669 | |||
XM_017007063.1 | XP_016862552.1 | 17, 18 | 2155 | |||
GenBank mRNA | AB098597.1 | - | 21, 22 | 2856 | ||
AF543840.1 | - | 4, 5 | 672 | |||
AK027052.1 | - | 6, 7 | 954 | |||
AK075220.1 | - | 12, 13 | 1593 | |||
AK127826.1 | - | 22, 23 | 2951 | |||
AK223599.1 | - | 22, 23 | 3002 | |||
AK314478.1 | - | 22, 23 | 2898 | |||
AL157482.1 | - | 4, 5 | 619 | |||
AY358146.1 | - | 15, 16 | 1844 | |||
BC033635.2 | - | 10, 11 | 1446 | |||
BC039297.1 | - | 22, 23 | 2862 | 5297602 | ||
BX648340.1 | - | 22, 23 | 2893 |