Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022767.3 | NP_073604.3 | 4 | 911 | ||
XM_005254966.1 | XP_005255023.1 | 4 | 1101 | |||
XM_005254967.1 | XP_005255024.1 | 4 | 1241 | |||
XM_006720645.3 | XP_006720708.2 | 4 | 873 | |||
XM_011521905.2 | XP_011520207.1 | 6 | 1265 | |||
XM_017022489.1 | XP_016877978.1 | 4 | 1888 | |||
XM_017022490.1 | XP_016877979.1 | 6 | 1231 | |||
GenBank mRNA | AB209323.1 | - | 2 | 2835 | ||
AF318330.1 | - | 4 | 841 | |||
AF327352.1 | - | 3 | 832 | |||
AK022546.1 | - | 3 | 445 | |||
BC005164.1 | - | 3 | 827 | 3531883 | ||
BC014407.1 | - | 3 | 446 | 4558021 | ||
BC020988.1 | - | 4 | 849 | 3852419 |