Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GINS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126129.1 | NP_001119601.1 | 1 | 222 | ||
NM_001126130.1 | NP_001119602.1 | 1 | 222 | |||
NM_022770.3 | NP_073607.2 | 1 | 222 | |||
GenBank mRNA | AK023974.1 | - | 1 | 128 | ||
AK315622.1 | - | 1 | 173 | |||
BC005879.1 | - | 1 | 97 | 3529058 | ||
BC014437.1 | - | 1 | 116 | 4870865 | ||
BP374281.1 | - | 1 | 165 | |||
CR457283.1 | - | 1 | 14 | |||
DA550725.1 | - | 1 | 222 | |||
KU178752.1 | - | 1 | 14 | |||
KU178753.1 | - | 1 | 14 | |||
KU178754.1 | - | 1 | 14 | |||
KU178755.1 | - | 1 | 14 |