Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130821.2 | NP_001124293.1 | 3, 4 | 341 | ||
NM_001130822.2 | NP_001124294.1 | 2, 3 | 233 | |||
NM_001287525.1 | NP_001274454.1 | 4, 5 | 383 | |||
NM_022777.3 | NP_073614.1 | 3, 4 | 310 | |||
GenBank mRNA | AK023287.1 | - | 4, 5 | 341 | ||
AK024179.1 | - | 3, 4 | 236 | |||
AK294653.1 | - | 3, 4 | 295 | |||
AK298888.1 | - | 2, 3 | 174 | |||
AU133515.1 | - | 4, 5 | 341 | |||
BC009823.2 | - | 3, 4 | 283 | 3946565 | ||
BC038668.1 | - | 2, 3 | 1020 | 5261016 | ||
BU597007.1 | - | 3, 4 | 318 | |||
CA396999.1 | - | 3, 4 | 299 | |||
DN993868.1 | - | 3, 4 | 319 |