Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130820.2 | NP_001124292.1 | 2 | 181 | ||
NM_001130821.2 | NP_001124293.1 | 2 | 212 | |||
NM_001287525.1 | NP_001274454.1 | 3 | 254 | |||
NM_022777.3 | NP_073614.1 | 2 | 181 | |||
GenBank mRNA | AK023287.1 | - | 3 | 212 | ||
AK024179.1 | - | 2 | 107 | |||
AK292254.1 | - | 2 | 130 | |||
AK294653.1 | - | 2 | 166 | |||
AU133515.1 | - | 3 | 212 | |||
BC004522.2 | - | 2 | 135 | 3939191 | ||
BC009823.2 | - | 2 | 154 | 3946565 | ||
BC038668.1 | - | 2 | 149 | 5261016 | ||
BU597007.1 | - | 2 | 189 | |||
CA396999.1 | - | 2 | 170 | |||
DN993868.1 | - | 2 | 190 |