Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022785.3 | NP_073622.2 | 30 | 4403 | ||
NM_198856.2 | NP_942153.1 | 28 | 4045 | |||
XM_005261704.2 | XP_005261761.1 | 30 | 4343 | |||
XM_005261705.3 | XP_005261762.1 | 29 | 4257 | |||
XM_011530316.1 | XP_011528618.1 | 31 | 4630 | |||
XM_011530317.2 | XP_011528619.1 | 29 | 4190 | |||
XM_011530318.2 | XP_011528620.1 | 29 | 4286 | |||
XM_011530319.2 | XP_011528621.1 | 28 | 3978 | |||
XM_011530320.2 | XP_011528622.1 | 31 | 4442 | |||
XM_011530321.2 | XP_011528623.1 | 28 | 3973 | |||
XM_011530322.2 | XP_011528624.1 | 29 | 4101 | |||
XM_011530323.2 | XP_011528625.1 | 26 | 3759 | |||
XM_011530325.2 | XP_011528627.1 | 24 | 3504 | |||
XM_011530327.2 | XP_011528629.1 | 21 | 3251 | |||
XM_011530329.1 | XP_011528631.1 | 17 | 2723 | |||
XM_017028910.1 | XP_016884399.1 | 31 | 4442 | |||
XM_017028911.1 | XP_016884400.1 | 31 | 4442 | |||
XM_017028912.1 | XP_016884401.1 | 27 | 4101 | |||
GenBank mRNA | AB073862.1 | - | 10 | 1597 | ||
AK027241.1 | - | 10 | 1601 | |||
AK225596.1 | - | 10 | 1602 | |||
AK225605.1 | - | 10 | 1602 | |||
AK292411.1 | - | 30 | 4343 | |||
AK292498.1 | - | 28 | 4022 | |||
AL355841.1 | - | 5 | 726 | |||
BC039315.1 | - | 28 | 3898 | 5297436 |