Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022823.2 | NP_073734.1 | 3 | 563 | ||
XM_005264499.3 | XP_005264556.1 | 3 | 607 | |||
GenBank mRNA | AK026015.1 | - | 3 | 523 | ||
AY358136.1 | - | 3 | 281 | |||
BC032725.1 | - | 2 | 194 | 5533159 |