Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB593133.1 | - | 2, 3 | 329 | ||
AK001022.1 | - | 2, 3 | 325 | |||
BC011967.1 | - | 2, 3 | 312 | 3866562 | ||
BC012136.1 | - | 2, 3 | 324 | 4558235 | ||
DB055557.1 | - | 2, 3 | 408 |