Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282805.1 | NP_001269734.1 | 7 | 1767 | ||
NM_001282806.1 | NP_001269735.1 | 4 | 1471 | |||
NM_001282807.1 | NP_001269736.1 | 4 | 1514 | |||
NM_022826.3 | NP_073737.1 | 5 | 1639 | |||
XM_005246773.2 | XP_005246830.1 | 5 | 1662 | |||
XM_005246774.1 | XP_005246831.1 | 5 | 1662 | |||
XM_017004718.1 | XP_016860207.1 | 5 | 1639 | |||
XM_017004719.1 | XP_016860208.1 | 4 | 1497 | |||
XM_017004720.1 | XP_016860209.1 | 4 | 1459 | |||
XM_017004721.1 | XP_016860210.1 | 4 | 1489 | |||
GenBank mRNA | AK316382.1 | - | 4 | 1471 | ||
AL597638.1 | - | 1 | 252 |