Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258372.1 | NP_001245301.1 | 16 | 2446 | ||
NM_001258373.1 | NP_001245302.1 | 17 | 2549 | |||
NM_022827.3 | NP_073738.2 | 17 | 2494 | |||
GenBank mRNA | AF086245.1 | - | 3 | 341 | ||
AK025000.1 | - | 16 | 2343 | |||
AK025622.1 | - | 12 | 1735 | |||
AK092731.1 | - | 17 | 2479 | |||
AK125807.1 | - | 18 | 3316 | |||
AK225189.1 | - | 16 | 2343 | |||
AK291805.1 | - | 17 | 2437 | |||
AL137367.1 | - | 2 | 249 | |||
BC017468.1 | - | 6 | 938 | |||
BC025255.1 | - | 17 | 2382 | 5019010 | ||
BC065526.1 | - | 17 | 2436 | 6149633 | ||
BC111029.1 | - | 16 | 2351 | 5245538 | ||
CR936769.1 | - | 14 | 2973 | |||
DQ238597.1 | - | 16 | 2444 | |||
GQ472220.1 | - | 16 | 2351 |