Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022830.2 | NP_073741.2 | 9 | 2759 | ||
GenBank mRNA | AB101208.1 | - | 9 | 2673 | ||
AK025920.1 | - | 5 | 1722 | |||
AK026000.1 | - | 9 | 2666 | |||
AK292610.1 | - | 9 | 2758 | |||
BC005013.1 | - | 5 | 1721 | |||
BC110910.1 | - | 9 | 2648 | 6141937 | ||
BC128263.1 | - | 9 | 2652 | 40118672 | ||
CA444886.1 | - | 1 | 82 | |||
KU178756.1 | - | 5 | 1590 | |||
KU178757.1 | - | 5 | 475 |