Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022835.2 | NP_073746.2 | 11 | 1520 | ||
XM_005259163.2 | XP_005259220.1 | 11 | 1341 | |||
XM_006723334.2 | XP_006723397.1 | 11 | 1871 | |||
XM_011527232.2 | XP_011525534.1 | 11 | 1447 | |||
XM_017027150.1 | XP_016882639.1 | 11 | 2051 | |||
XM_017027151.1 | XP_016882640.1 | 11 | 2051 | |||
GenBank mRNA | AK024429.1 | - | 11 | 1520 | ||
AK026111.1 | - | 11 | 1329 | |||
AK302476.1 | - | 11 | 1144 | |||
BC013426.1 | - | 11 | ||||
BC075858.1 | - | 9 | 885 |