Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCLRE1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319946.1 | NP_001306875.1 | 3 | 851 | ||
NM_001319947.1 | NP_001306876.1 | 4 | 765 | |||
NM_022836.3 | NP_073747.1 | 4 | 1017 | |||
GenBank mRNA | AF306691.1 | - | 3 | 735 | ||
AF306692.1 | - | 3 | 392 | |||
AF306693.1 | - | 4 | 952 | |||
AK022872.1 | - | 4 | 563 | |||
AK024060.1 | - | 4 | 935 | |||
BC029687.1 | - | 4 | 894 | 5001181 | ||
CR936679.1 | - | 3 | 776 |