Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032014.2 | NP_114403.1 | 4 | 474 | ||
GenBank mRNA | AF161453.1 | - | 4 | 401 | ||
BC012167.1 | - | 4 | 426 | 4637888 | ||
CA413186.1 | - | 1 | 236 |
Gene Symbol: URGCP-MRPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204871.1 | NP_001191800.1 | 7 | 616 |