Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031280.3 | NP_112570.2 | 8 | 902 | ||
GenBank mRNA | AB049946.1 | - | 8 | 777 | ||
AB209905.1 | - | 6 | 5055 | |||
AF265439.1 | - | 8 | 821 | |||
AK021626.1 | - | 4 | 1562 | |||
AK125230.1 | - | 3 | 2076 | |||
AK315441.1 | - | 8 | 841 | |||
BC031336.1 | - | 8 | 817 | 3543896 | ||
CF123360.1 | - | 2 | 307 |