Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDRG4 SIRNA Assays ›
Gene Symbol:
NDRG4
Gene Name:
NDRG family member 4
Gene Aliases:
BDM1, SMAP-8, SMAP8
Chromosome Location:
Chr.16: 58463645 - 58515403 on Build GRCh38
UniGene ID:
Hs.322430
Species:
Human
siRNA ID
HSS127940
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDRG4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130487.1
NP_001123959.1
8
928
NM_001242833.1
NP_001229762.1
7
627
NM_001242834.1
NP_001229763.1
6
591
NM_001242835.1
NP_001229764.1
6
537
NM_001242836.1
NP_001229765.1
6
537
NM_020465.3
NP_065198.1
8
703
NM_022910.3
NP_075061.1
8
800
NR_040072.1
6
629
XM_006721252.3
XP_006721315.1
9
803
XM_006721253.3
XP_006721316.1
9
736
XM_006721254.3
XP_006721317.1
8
790
XM_006721256.3
XP_006721319.1
9
734
XM_006721257.3
XP_006721320.1
8
775
XM_006721258.3
XP_006721321.1
8
679
XM_006721259.3
XP_006721322.1
8
744
XM_006721260.3
XP_006721323.1
8
682
XM_006721261.3
XP_006721324.1
7
627
XM_006721262.3
XP_006721325.1
6
708
XM_011523290.2
XP_011521592.1
9
890
XM_011523291.2
XP_011521593.1
8
854
XM_011523292.2
XP_011521594.1
9
890
XM_011523293.2
XP_011521595.1
8
848
XM_011523294.2
XP_011521596.1
9
669
XM_017023582.1
XP_016879071.1
8
854
XM_017023583.1
XP_016879072.1
9
890
XM_017023584.1
XP_016879073.1
9
803
XM_017023585.1
XP_016879074.1
8
790
XM_017023586.1
XP_016879075.1
8
737
XM_017023587.1
XP_016879076.1
9
734
XM_017023588.1
XP_016879077.1
8
679
XM_017023589.1
XP_016879078.1
8
744
XM_017023590.1
XP_016879079.1
9
669
GenBank mRNA
AB021172.1
-
8
603
AB033006.2
-
6
523
AB044944.1
-
6
537
AB044945.1
-
6
537
AB044946.1
-
7
550
AF308608.1
-
6
544
AK055038.1
-
6
539
AK124005.1
-
2
506
AK126574.1
-
8
700
AK126729.1
-
9
2650
AK131248.1
-
6
591
AK296415.1
-
6
591
AK299949.1
-
7
627
AK316089.1
-
6
629
AL136584.1
-
6
523
BC001527.1
-
6
521
BC011795.2
-
6
525
4139594
CR936733.1
-
8
614
KU178760.1
-
6
431
KU178761.1
-
6
431
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
heart looping
response to stress
brain development
visual learning
negative regulation of platelet-derived growth factor receptor signaling pathway
positive regulation of neuron projection development
negative regulation of smooth muscle cell migration
cell growth
cell differentiation
embryonic heart tube development
vesicle docking
negative regulation of smooth muscle cell proliferation
cardiac muscle cell proliferation
cell migration involved in heart development
positive regulation of ERK1 and ERK2 cascade
regulation of endocytic recycling
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline