Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133489.2 | NP_597996.2 | 12 | 1724 | ||
GenBank mRNA | AF331523.1 | - | 12 | 1724 | ||
AK122981.1 | - | 13 | 1392 | |||
AK314085.1 | - | 3 | 1150 | |||
AL050358.1 | - | 6 | 2561 |
Gene Symbol: B4GALNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268773.4 | XP_005268830.1 | 11 | 3534 | ||
XM_011538147.2 | XP_011536449.1 | 12 | 3552 | |||
XM_011538148.2 | XP_011536450.1 | 12 | 3519 | |||
XM_017019140.1 | XP_016874629.1 | 11 | 3636 | |||
XM_017019141.1 | XP_016874630.1 | 11 | 3348 | |||
XM_017019142.1 | XP_016874631.1 | 11 | 3213 | |||
GenBank mRNA | AB209460.1 | - | 10 | 3716 |