Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133489.2 | NP_597996.2 | 14 | 1892 | ||
GenBank mRNA | AF331523.1 | - | 14 | 1892 | ||
AK122981.1 | - | 15 | 1548 | |||
AK314085.1 | - | 5 | 1306 | |||
AL050358.1 | - | 8 | 2717 |
Gene Symbol: B4GALNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268773.4 | XP_005268830.1 | 11 | 3170 | ||
XM_011538147.2 | XP_011536449.1 | 12 | 3188 | |||
XM_011538148.2 | XP_011536450.1 | 12 | 3155 | |||
XM_017019140.1 | XP_016874629.1 | 11 | 3272 | |||
XM_017019141.1 | XP_016874630.1 | 11 | 2984 | |||
XM_017019142.1 | XP_016874631.1 | 11 | 2849 | |||
GenBank mRNA | AB209460.1 | - | 10 | 3352 |