Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203365.3 | NP_976241.1 | 8 | 1333 | ||
NM_213589.2 | NP_998754.1 | 6 | 1177 | |||
XM_005246777.3 | XP_005246834.1 | 8 | 1328 | |||
XM_005246779.4 | XP_005246836.1 | 8 | 2227 | |||
XM_006712695.3 | XP_006712758.1 | 9 | 2308 | |||
XM_011511646.2 | XP_011509948.1 | 9 | 1400 | |||
XM_011511647.1 | XP_011509949.1 | 6 | 1033 | |||
XM_011511648.1 | XP_011509950.1 | 6 | 1027 | |||
XM_011511649.2 | XP_011509951.1 | 8 | 1328 | |||
GenBank mRNA | AB051468.1 | - | 5 | 910 | ||
AB053311.1 | - | 8 | 1327 | |||
AJ584699.1 | - | 6 | 1171 | |||
AY494951.1 | - | 6 | 1173 | |||
AY523977.1 | - | 8 | 1329 | |||
AY523978.1 | - | 8 | 1329 |