Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044385.2 | NP_001037850.1 | 6 | 490 | ||
NM_152388.3 | NP_689601.2 | 6 | 655 | |||
GenBank mRNA | AB053301.1 | - | 6 | 657 | ||
AF212043.1 | - | 1 | 59 | |||
AK090601.1 | - | 6 | 348 | |||
AK303954.1 | - | 6 | 462 | |||
BQ219059.1 | - | 6 | 462 |