Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044385.2 | NP_001037850.1 | 8, 9 | 807 | ||
NM_152388.3 | NP_689601.2 | 8, 9 | 972 | |||
GenBank mRNA | AB053301.1 | - | 8, 9 | 974 | ||
AF212043.1 | - | 3, 4 | 376 | |||
AK090601.1 | - | 8, 9 | 665 | |||
AK303954.1 | - | 8, 9 | 794 | |||
AK304395.1 | - | 6, 7 | 496 | |||
BC029611.1 | - | 8, 9 | 720 | 5304046 |