Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 35, 36 | 5955 | ||
XM_005246787.3 | XP_005246844.1 | 35, 36 | 5981 | |||
XM_005246788.2 | XP_005246845.1 | 14, 15 | 2457 | |||
XM_006712698.3 | XP_006712761.1 | 36, 37 | 6068 | |||
XM_006712699.3 | XP_006712762.1 | 36, 37 | 6068 | |||
XM_011511658.2 | XP_011509960.1 | 36, 37 | 6023 | |||
XM_011511659.2 | XP_011509961.1 | 36, 37 | 6080 | |||
XM_011511660.2 | XP_011509962.1 | 32, 33 | 5402 | |||
XM_011511662.2 | XP_011509964.1 | 28, 29 | 4737 | |||
XM_011511663.2 | XP_011509965.1 | 36, 37 | 6058 | |||
XM_017004728.1 | XP_016860217.1 | 23, 24 | 4135 | |||
XM_017004729.1 | XP_016860218.1 | 11, 12 | 2149 | |||
GenBank mRNA | AK127772.1 | - | 9, 10 | 1823 | ||
AK128636.1 | - | 5, 6 | 682 | |||
AY172970.1 | - | 5, 6 | 861 |