Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS33A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022916.4 | NP_075067.2 | 6 | 810 | ||
GenBank mRNA | AF439857.1 | - | 6 | 721 | ||
AK026840.1 | - | 6 | 720 | |||
AK315545.1 | - | 6 | 748 | |||
BC016617.1 | - | 6 | 718 | 3855313 |