Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022917.4 | NP_075068.2 | 10 | 1342 | ||
NM_139235.3 | NP_631981.2 | 10 | 1342 | |||
XM_017015044.1 | XP_016870533.1 | 10 | 1278 | |||
GenBank mRNA | AF361079.1 | - | 10 | 1300 | ||
AF361080.1 | - | 10 | 1300 | |||
AF361081.1 | - | 10 | 1300 | |||
AK056559.1 | - | 2 | 56 | |||
AK293969.1 | - | 9 | 1121 | |||
BC030139.2 | - | 10 | 1246 | 4423152 |