Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022917.4 | NP_075068.2 | 14, 15 | 1981 | ||
NM_139235.3 | NP_631981.2 | 14, 15 | 1981 | |||
XM_017015044.1 | XP_016870533.1 | 14, 15 | 1937 | |||
GenBank mRNA | AF361079.1 | - | 14, 15 | 1939 | ||
AF361080.1 | - | 14, 15 | 1939 | |||
AF361081.1 | - | 14, 15 | 1939 | |||
AK056559.1 | - | 6, 7 | 695 | |||
AK293969.1 | - | 13, 14 | 1784 | |||
BC008852.2 | - | 3, 4 | 374 | |||
BC030139.2 | - | 14, 15 | 1885 | 4423152 |