Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023011.3 | NP_075387.1 | 3 | 390 | ||
NM_080687.2 | NP_542418.1 | 3 | 390 | |||
XM_006719991.3 | XP_006720054.1 | 2 | 222 | |||
XM_011534844.1 | XP_011533146.1 | 3 | 368 | |||
XM_011534845.2 | XP_011533147.1 | 3 | 875 | |||
XM_011534846.1 | XP_011533148.1 | 3 | 337 | |||
XM_011534847.2 | XP_011533149.1 | 1 | 465 | |||
XM_011534848.2 | XP_011533150.1 | 3 | 875 | |||
XM_017020709.1 | XP_016876198.1 | 2 | 1059 | |||
XM_017020710.1 | XP_016876199.1 | 2 | 222 | |||
XM_017020711.1 | XP_016876200.1 | 2 | 222 | |||
XM_017020712.1 | XP_016876201.1 | 3 | 383 | |||
GenBank mRNA | AF318575.1 | - | 3 | 283 | ||
AK294545.1 | - | 3 | 337 | |||
AK302003.1 | - | 3 | 326 | |||
AY013250.1 | - | 3 | 390 | |||
BC023569.1 | - | 3 | 354 | 4127465 |