Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324475.1 | NP_001311404.1 | 17 | 1883 | ||
NM_023015.4 | NP_075391.3 | 16 | 2224 | |||
XM_005245459.2 | XP_005245516.1 | 16 | 2241 | |||
XM_005245461.2 | XP_005245518.1 | 10 | 914 | |||
XM_006711490.3 | XP_006711553.1 | 17 | 1694 | |||
XM_011509906.2 | XP_011508208.1 | 16 | 1606 | |||
XM_017002132.1 | XP_016857621.1 | 6 | 482 | |||
GenBank mRNA | AK025572.1 | - | 2 | 45 | ||
AK074979.1 | - | 9 | 922 | |||
AK225294.1 | - | 2 | 45 | |||
AK290025.1 | - | 17 | 1877 | |||
AK298746.1 | - | 4 | 374 | |||
AK304874.1 | - | 10 | 1313 | |||
AL080172.1 | - | 11 | 1123 | |||
AL832133.1 | - | 15 | 3259 | |||
BC054513.1 | - | 3 | 260 | |||
BC098431.1 | - | 2 | 84 | |||
BC105092.1 | - | 16 | 1687 | 8144095 | ||
BC105094.1 | - | 16 | 1687 | 8144097 | ||
BC116458.1 | - | 16 | 2219 | 5752962 | ||
BX640786.1 | - | 16 | 2210 | |||
BX640950.1 | - | 16 | 2225 | |||
CR749212.1 | - | 14 | 3751 | |||
CR749376.1 | - | 15 | 3876 |