Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293285.1 | NP_001280214.1 | 15 | 2682 | ||
NM_001293286.1 | NP_001280215.1 | 13 | 2535 | |||
NM_023071.3 | NP_075559.2 | 14 | 2598 | |||
NR_120663.1 | 15 | 2642 | ||||
XM_011538690.2 | XP_011536992.1 | 15 | 2070 | |||
XM_011538691.2 | XP_011536993.1 | 13 | 1965 | |||
XM_017019840.1 | XP_016875329.1 | 14 | 1904 | |||
GenBank mRNA | AB075515.1 | - | 12 | 1626 | ||
AK023179.1 | - | 10 | 1507 | |||
AK023202.1 | - | 14 | 1997 | |||
AK024678.1 | - | 6 | 894 | |||
AK095582.1 | - | 15 | 2642 | |||
AK292334.1 | - | 13 | 1935 | |||
AL833614.1 | - | 15 | 2123 | |||
BC048299.1 | - | 13 | 1965 | 5520818 | ||
BX648916.1 | - | 13 | 1908 |