Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321299.1 | NP_001308228.1 | 6 | 1473 | ||
NM_032387.4 | NP_115763.2 | 7 | 1562 | |||
XM_011525135.2 | XP_011523437.1 | 7 | 2492 | |||
XM_017024962.1 | XP_016880451.1 | 7 | 2492 | |||
XM_017024963.1 | XP_016880452.1 | 7 | 2492 | |||
XM_017024964.1 | XP_016880453.1 | 7 | 2492 | |||
XM_017024965.1 | XP_016880454.1 | 7 | 2492 | |||
XM_017024966.1 | XP_016880455.1 | 4 | 614 | |||
GenBank mRNA | AF390018.1 | - | 7 | 1515 | ||
AJ316534.1 | - | 7 | 1511 | |||
AK096052.1 | - | 5 | 1031 | |||
AK301963.1 | - | 2 | 174 | |||
BC136664.1 | - | 7 | 1512 | 9020654 |