Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002838.3 | NP_001002838.1 | 3, 4 | 1143 | ||
NM_020922.4 | NP_065973.2 | 3, 4 | 1143 | |||
XM_006724593.3 | XP_006724656.1 | 3, 4 | 1081 | |||
XM_011530802.2 | XP_011529104.1 | 3, 4 | 1081 | |||
XM_017029741.1 | XP_016885230.1 | 3, 4 | 1082 | |||
XM_017029742.1 | XP_016885231.1 | 3, 4 | 1159 | |||
XM_017029743.1 | XP_016885232.1 | 3, 4 | 1074 | |||
XM_017029744.1 | XP_016885233.1 | 3, 4 | 976 | |||
XM_017029745.1 | XP_016885234.1 | 3, 4 | 1081 | |||
GenBank mRNA | AJ409088.2 | - | 2, 3 | 704 | ||
AY082340.1 | - | 2, 3 | 704 | |||
AY352048.1 | - | 3, 4 | 1143 |