Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009877.2 | NP_001009877.2 | 7 | 769 | ||
NM_001317951.1 | NP_001304880.1 | 7 | 1121 | |||
NM_023924.4 | NP_076413.3 | 7 | 942 | |||
NR_134296.1 | 7 | 1278 | ||||
XM_006714494.3 | XP_006714557.1 | 7 | 981 | |||
XM_017009780.1 | XP_016865269.1 | 7 | 981 | |||
GenBank mRNA | AK023503.1 | - | 7 | 1099 | ||
AK024392.1 | - | 4 | 777 | |||
AK026830.1 | - | 2 | 73 | |||
AK297573.1 | - | 7 | 756 | |||
AK299157.1 | - | 7 | 942 | |||
AK301987.1 | - | 7 | 893 | |||
BC007217.1 | - | 7 | 958 | |||
BC041590.1 | - | 5 | 628 | 5428011 | ||
BX648703.1 | - | 7 | 1280 | |||
DQ248311.1 | - | 3 | 267 |