Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146319.1 | NP_001139791.1 | 9, 10 | 1117 | ||
NM_001146320.1 | NP_001139792.1 | 8, 9 | 1154 | |||
NM_001146321.1 | NP_001139793.1 | 10, 11 | 1309 | |||
NM_001146322.1 | NP_001139794.1 | 9, 10 | 928 | |||
NM_023927.2 | NP_076416.2 | 9, 10 | 1266 | |||
XM_005272058.3 | XP_005272115.1 | 9, 10 | 1004 | |||
XM_005272063.3 | XP_005272120.1 | 9, 10 | 1201 | |||
XM_011543592.2 | XP_011541894.1 | 10, 11 | 1978 | |||
XM_017009782.1 | XP_016865271.1 | 10, 11 | 1111 | |||
XM_017009783.1 | XP_016865272.1 | 8, 9 | 722 | |||
XM_017009785.1 | XP_016865274.1 | 8, 9 | 963 | |||
GenBank mRNA | AK024966.1 | - | 9, 10 | 1034 | ||
AK225829.1 | - | 9, 10 | 1034 | |||
AK293307.1 | - | 8, 9 | 859 | |||
AK296267.1 | - | 10, 11 | 1014 | |||
AK298009.1 | - | 9, 10 | 851 | |||
AK298349.1 | - | 9, 10 | 939 | |||
AK300154.1 | - | 9, 10 | 1062 | |||
AK303857.1 | - | 9, 10 | 868 | |||
AK316288.1 | - | 9, 10 | 1081 | |||
AY116970.1 | - | 9, 10 | 836 | |||
BC008590.1 | - | 9, 10 | 999 | 4148772 |