Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142674.1 | NP_001136146.1 | 10 | 1034 | ||
NM_001142675.1 | NP_001136147.1 | 9 | 1144 | |||
NM_001142676.1 | NP_001136148.1 | 10 | 1005 | |||
NM_001142677.1 | NP_001136149.1 | 8 | 837 | |||
NM_023947.3 | NP_076436.3 | 9 | 930 | |||
XM_005253072.2 | XP_005253129.1 | 9 | 1017 | |||
XM_005253073.4 | XP_005253130.1 | 9 | 925 | |||
XM_006718282.2 | XP_006718345.1 | 10 | 1005 | |||
XM_011520299.1 | XP_011518601.1 | 9 | 1241 | |||
XM_011520300.1 | XP_011518602.1 | 8 | 1148 | |||
XM_011520301.2 | XP_011518603.1 | 9 | 1241 | |||
XM_011520302.2 | XP_011518604.1 | 9 | 1241 | |||
XM_017018166.1 | XP_016873655.1 | 10 | 1091 | |||
XM_017018167.1 | XP_016873656.1 | 9 | 896 | |||
XM_017018168.1 | XP_016873657.1 | 10 | 1017 | |||
GenBank mRNA | AF212229.1 | - | 10 | 968 | ||
AK027711.1 | - | 9 | 879 | |||
AK075413.1 | - | 10 | 941 | |||
AK124697.1 | - | 9 | 1506 | |||
AK225025.1 | - | 10 | 970 | |||
AK225398.1 | - | 10 | 953 | |||
AK297743.1 | - | 9 | 1011 | |||
AK301245.1 | - | 9 | 990 | |||
AY037151.1 | - | 8 | 798 | |||
BC000001.2 | - | 10 | 954 | 3504261 | ||
BC013642.1 | - | 7 | 566 | |||
BC095409.1 | - | 9 | 866 | 30333427 |