Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256737.1 | NP_001243666.1 | 4 | 752 | ||
NM_001256738.1 | NP_001243667.1 | 4 | 542 | |||
NM_001256739.1 | NP_001243668.1 | 3 | 521 | |||
NM_024034.4 | NP_076939.3 | 4 | 695 | |||
NR_046353.1 | 2 | 328 | ||||
GenBank mRNA | AF277566.1 | - | 4 | 650 | ||
AK056251.1 | - | 4 | 685 | |||
AK225166.1 | - | 4 | 685 | |||
AK293484.1 | - | 3 | 471 | |||
AK297348.1 | - | 2 | 230 | |||
AK299259.1 | - | 5 | 607 | |||
AK299700.1 | - | 4 | 701 | |||
BC000199.1 | - | 4 | 671 | 3352559 | ||
BC009014.1 | - | 4 | 634 | 4155653 |