Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287490.1 | NP_001274419.1 | 3 | 638 | ||
NM_024037.2 | NP_076942.1 | 3 | 638 | |||
XM_017002300.1 | XP_016857789.1 | 3 | 709 | |||
GenBank mRNA | AK024326.1 | - | 3 | 537 | ||
AK223419.1 | - | 3 | 574 | |||
BC000209.1 | - | 3 | 592 | 3350471 | ||
GQ844687.1 | - | 3 | 592 |