Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM106C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143841.1 | NP_001137313.1 | 4, 5 | 524 | ||
NM_001143842.1 | NP_001137314.1 | 4, 5 | 595 | |||
NM_001143843.1 | NP_001137315.1 | 4, 5 | 595 | |||
NM_024056.3 | NP_076961.1 | 4, 5 | 524 | |||
GenBank mRNA | AK299154.1 | - | 4, 5 | 464 | ||
AK313698.1 | - | 4, 5 | 466 | |||
BC000854.2 | - | 3, 4 | 437 | 3459189 | ||
BC107792.1 | - | 4, 5 | 467 | 30321820 | ||
BP228371.1 | - | 2, 3 | 181 | |||
BQ227924.1 | - | 4, 5 | 466 | |||
CN256547.1 | - | 4, 5 | 506 | |||
CX763149.1 | - | 4, 5 | 508 | |||
DA567999.1 | - | 3, 4 | 251 | |||
DA571733.1 | - | 4, 5 | 524 | |||
DQ020163.1 | - | 3, 4 | 409 |