Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111322.1 | NP_001104792.1 | 18, 19 | 2336 | ||
NM_024072.3 | NP_076977.3 | 18, 19 | 2336 | |||
GenBank mRNA | AY148094.1 | - | 18, 19 | 2315 | ||
BC005848.2 | - | 18, 19 | 2257 |