Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SECISBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282688.1 | NP_001269617.1 | 5 | 767 | ||
NM_001282689.1 | NP_001269618.1 | 5 | 649 | |||
NM_001282690.1 | NP_001269619.1 | 5 | 847 | |||
NM_024077.4 | NP_076982.3 | 5 | 770 | |||
XM_005252196.2 | XP_005252253.1 | 5 | 737 | |||
XM_006717282.2 | XP_006717345.1 | 5 | 703 | |||
XM_011519000.2 | XP_011517302.1 | 5 | 824 | |||
XM_011519001.1 | XP_011517303.1 | 5 | 824 | |||
XM_011519002.1 | XP_011517304.1 | 5 | 824 | |||
XM_011519003.1 | XP_011517305.1 | 5 | 824 | |||
XM_017015122.1 | XP_016870611.1 | 5 | 737 | |||
XM_017015123.1 | XP_016870612.1 | 5 | 737 | |||
XM_017015124.1 | XP_016870613.1 | 5 | 737 | |||
XM_017015125.1 | XP_016870614.1 | 3 | 363 | |||
GenBank mRNA | AB208940.1 | - | 5 | 728 | ||
AF380995.1 | - | 5 | 723 | |||
AK090608.1 | - | 5 | 737 | |||
AK290182.1 | - | 5 | 616 | |||
AK299161.1 | - | 5 | 858 | |||
AK302852.1 | - | 5 | 765 | |||
AL136881.1 | - | 5 | 603 | |||
BC001189.1 | - | 5 | 603 | |||
BC036109.1 | - | 5 | 732 | 5296292 | ||
BC068466.1 | - | 5 | 705 | |||
BC099900.1 | - | 5 | 706 | |||
BX648787.1 | - | 3 | 3710 |