Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SECISBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282688.1 | NP_001269617.1 | 1, 2 | 139 | ||
NM_001282689.1 | NP_001269618.1 | 1, 2 | 139 | |||
NM_024077.4 | NP_076982.3 | 1, 2 | 139 | |||
XM_005252196.2 | XP_005252253.1 | 1, 2 | 106 | |||
XM_017015122.1 | XP_016870611.1 | 1, 2 | 106 | |||
XM_017015123.1 | XP_016870612.1 | 1, 2 | 106 | |||
XM_017015124.1 | XP_016870613.1 | 1, 2 | 106 | |||
GenBank mRNA | AB208940.1 | - | 1, 2 | 97 | ||
AF380995.1 | - | 1, 2 | 92 | |||
AK090608.1 | - | 1, 2 | 106 | |||
AK290182.1 | - | 1, 2 | 106 | |||
AK302852.1 | - | 1, 2 | 137 | |||
AL136881.1 | - | 1, 2 | 93 | |||
BC001189.1 | - | 1, 2 | 93 | |||
BC036109.1 | - | 1, 2 | 101 | 5296292 | ||
BC068466.1 | - | 1, 2 | 74 | |||
BC099900.1 | - | 1, 2 | 75 | |||
BP219148.1 | - | 1, 2 | 139 | |||
DR002073.1 | - | 1, 2 | 132 |