Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077198.2 | NP_001070666.1 | 8 | 1324 | ||
NM_024085.4 | NP_076990.4 | 7 | 1272 | |||
NR_104255.1 | 8 | 1324 | ||||
GenBank mRNA | AK025822.1 | - | 2 | 626 | ||
AK056595.1 | - | 8 | 1265 | |||
AK289755.1 | - | 8 | 1219 | |||
AK302521.1 | - | 7 | 1163 | |||
AK315553.1 | - | 2 | 636 | |||
AL833865.1 | - | 1 | 521 | |||
BC065534.1 | - | 8 | 1262 | 5787445 | ||
BX537984.1 | - | 7 | 1193 | |||
BX538192.1 | - | 7 | 1167 | |||
BX538198.1 | - | 8 | 1101 |