Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077198.2 | NP_001070666.1 | 6 | 527 | ||
NM_024085.4 | NP_076990.4 | 5 | 475 | |||
NR_104255.1 | 6 | 527 | ||||
GenBank mRNA | AK056595.1 | - | 6 | 468 | ||
AK289755.1 | - | 6 | 422 | |||
AK302521.1 | - | 5 | 366 | |||
BC065534.1 | - | 6 | 465 | 5787445 | ||
BP284912.1 | - | 2 | 185 | |||
BX537984.1 | - | 5 | 396 | |||
BX538192.1 | - | 5 | 370 | |||
BX538198.1 | - | 6 | 304 | |||
DC347583.1 | - | 5 | 475 |